Assay Details
Gene Symbol: MSANTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044657.1 | XP_017900146.1 | 3 - 4 | 831 | 78 | |
GenBank mRNA | JO445911.2 | - | 2 - 3 | 397 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
