Assay Details
Gene Symbol: FAM49A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055616.1 | XP_017911105.1 | 8 - 9 | 868 | 63 | |
XM_018055617.1 | XP_017911106.1 | 8 - 9 | 954 | 63 | ||
GenBank mRNA | JO578801.1 | - | 8 - 9 | 944 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
