Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059322.1 | XP_017914811.1 | 2 - 3 | 517 | 98 | |
XM_018059326.1 | XP_017914815.1 | 2 - 3 | 517 | 98 | ||
GenBank mRNA | JT724187.1 | - | 8 - 9 | 2474 | 98 |
Gene Symbol: LOC108637769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060239.1 | XP_017915728.1 | 1 - 2 | 472 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
