Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059322.1 | XP_017914811.1 | 3 - 4 | 627 | 125 | |
XM_018059323.1 | XP_017914812.1 | 4 - 5 | 379 | 125 | ||
XM_018059326.1 | XP_017914815.1 | 3 - 4 | 627 | 183 | ||
GenBank mRNA | JT724187.1 | - | 7 - 8 | 2364 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018059323.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
