Assay Details
Gene Symbol: GAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061762.1 | XP_017917251.1 | 3 - 4 | 810 | 65 | |
GenBank mRNA | KA342010.1 | - | 3 - 4 | 750 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
