Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973392.2 | XP_013828846.2 | 1 - 2 | 1778 | 71 | |
| XM_013973393.2 | XP_013828847.2 | 1 - 2 | 1778 | 71 | ||
More Information
Set Membership: |
|

MYD88
Genomic Map