Assay Details
| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681653.3 | XP_005681710.2 | 8 - 9 | 1700 | 71 | |
| XM_018049507.1 | XP_017904996.1 | 8 - 9 | 1569 | 71 | ||
More Information
Set Membership: |
|

CENPC
Genomic Map