Assay Details
| Gene Symbol: RNF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049831.1 | XP_017905320.1 | 7 - 8 | 706 | 67 | |
| XM_018049832.1 | XP_017905321.1 | 7 - 8 | 666 | 67 | ||
| XM_018049833.1 | XP_017905322.1 | 7 - 8 | 648 | 67 | ||
| GenBank mRNA | JO410723.2 | - | 7 - 8 | 685 | 67 | |
| JO541078.2 | - | 7 - 8 | 670 | 67 | ||
More Information
Set Membership: |
|

RNF4
Genomic Map