Assay Details
| Gene Symbol: SLC22A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682678.3 | XP_005682735.3 | 9 - 10 | 1835 | 69 | |
| GenBank mRNA | JO426245.1 | - | 1 - 2 | 1476 | 69 | |
More Information
Set Membership: |
|

SLC22A5
Genomic Map