Assay Details
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055764.1 | XP_017911253.1 | 3 - 4 | 255 | 83 | |
XM_018055765.1 | XP_017911254.1 | 3 - 4 | 255 | 83 | ||
XM_018055766.1 | XP_017911255.1 | 3 - 4 | 255 | 83 | ||
GenBank mRNA | JO588282.2 | - | 3 - 4 | 250 | 83 | |
JO647532.2 | - | 5 - 6 | 913 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
