Assay Details
| Gene Symbol: SEC14L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691630.2 | XP_005691687.2 | 1 - 2 | 138 | 65 | |
| GenBank mRNA | JO560426.2 | - | 11 - 12 | 1397 | 65 | |
More Information
Set Membership: |
|

SEC14L3
Genomic Map