Assay Details
| Gene Symbol: PLA2G15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692174.2 | XP_005692231.1 | 2 - 3 | 392 | 62 | |
| GenBank mRNA | JO325743.2 | - | 4 - 5 | 1655 | 62 | |
More Information
Set Membership: |
|

PLA2G15
Genomic Map