Assay Details
Gene Symbol: PRR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693110.2 | XP_005693167.2 | 10 - 11 | 1194 | 60 | |
XM_005693111.3 | XP_005693168.2 | 9 - 10 | 1101 | 60 | ||
XM_013971791.2 | XP_013827245.2 | 10 - 11 | 1328 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
