Assay Details
| Gene Symbol: ZKSCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697622.3 | XP_005697679.2 | 3 - 4 | 1445 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZKSCAN2| Gene Symbol: ZKSCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697622.3 | XP_005697679.2 | 3 - 4 | 1445 | 59 | |
Set Membership: |
|