Assay Details
Gene Symbol: LOC102183750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697657.3 | XP_005697714.1 | 7 - 8 | 1298 | 81 | |
XM_018040158.1 | XP_017895647.1 | 7 - 8 | 1337 | 81 | ||
XM_018040159.1 | XP_017895648.1 | 7 - 8 | 1297 | 81 | ||
XM_018040161.1 | XP_017895650.1 | 6 - 7 | 1090 | 81 | ||
XM_018040162.1 | XP_017895651.1 | 6 - 7 | 1127 | 81 | ||
XM_018040163.1 | XP_017895652.1 | 6 - 7 | 1087 | 81 | ||
GenBank mRNA | JO582012.1 | - | 7 - 8 | 1338 | 81 | |
JO582013.1 | - | 7 - 8 | 1374 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
