Assay Details
| Gene Symbol: SOX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048743.1 | XP_017904232.1 | 3 - 4 | 1014 | 92 | |
| GenBank mRNA | JO633129.2 | - | 2 - 3 | 945 | 92 | |
| JT763248.1 | - | 2 - 3 | 715 | 92 | ||
More Information
Set Membership: |
|

SOX10
Genomic Map