Assay Details
| Gene Symbol: SYNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048786.1 | XP_017904275.1 | 3 - 4 | 562 | 72 | |
| XM_018048787.1 | XP_017904276.1 | 3 - 4 | 582 | 72 | ||
| XM_018048788.1 | XP_017904277.1 | 3 - 4 | 511 | 72 | ||
More Information
Set Membership: |
|

SYNGR1
Genomic Map