Assay Details
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058499.1 | XP_017913988.1 | 10 - 11 | 1152 | 64 | |
XM_018058500.1 | XP_017913989.1 | 10 - 11 | 1462 | 64 | ||
XM_018058501.1 | XP_017913990.1 | 10 - 11 | 1089 | 64 | ||
XM_018058502.1 | XP_017913991.1 | 10 - 11 | 1245 | 64 | ||
GenBank mRNA | JO580542.1 | - | 5 - 6 | 2130 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
