Assay Details
Gene Symbol: PAX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054740.1 | XP_017910229.1 | 5 - 6 | 1403 | 85 | |
XM_018054746.1 | XP_017910235.1 | 5 - 6 | 1397 | 85 | ||
GenBank mRNA | JO545036.2 | - | 2 - 3 | 245 | 85 | |
JO545037.2 | - | 2 - 3 | 245 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
