Assay Details
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972565.2 | XP_013828019.1 | 6 - 7 | 882 | 71 | |
XM_013972566.2 | XP_013828020.1 | 6 - 7 | 879 | 71 | ||
GenBank mRNA | JO354141.2 | - | 1 - 2 | 265 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
