Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066537.1 | XP_017922026.1 | 2 - 3 | 554 | 79 | |
| GenBank mRNA | JO361312.2 | - | 2 - 3 | 566 | 79 | |
| JT749746.1 | - | 2 - 3 | 820 | 79 | ||
More Information
Set Membership: |
|

TMED3
Genomic Map