Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066915.1 | XP_017922404.1 | 9 - 10 | 941 | 60 | |
| XM_018066916.1 | XP_017922405.1 | 9 - 10 | 937 | 60 | ||
| XM_018066917.1 | XP_017922406.1 | 8 - 9 | 852 | 60 | ||
| XM_018066918.1 | XP_017922407.1 | 7 - 8 | 762 | 60 | ||
| XM_018066919.1 | XP_017922408.1 | 8 - 9 | 851 | 60 | ||
| XM_018066920.1 | XP_017922409.1 | 9 - 10 | 942 | 60 | ||
| XM_018066921.1 | XP_017922410.1 | 9 - 10 | 942 | 60 | ||
| GenBank mRNA | JO578459.1 | - | 6 - 7 | 1292 | 60 | |
| JO633288.2 | - | 6 - 7 | 1262 | 60 | ||
| KA347381.1 | - | 1050 | 60 | |||
More Information
Set Membership: |
|

PCBP4
Genomic Map