Assay Details
| Gene Symbol: RPL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674869.3 | XP_005674926.1 | 2 - 3 | 164 | 89 | |
| GenBank mRNA | JO341311.2 | - | 2 - 3 | 133 | 89 | |
| JO362713.2 | - | 2 - 3 | 116 | 89 | ||
| JO377262.2 | - | 2 - 3 | 126 | 89 | ||
| JO421375.2 | - | 2 - 3 | 158 | 89 | ||
| JO453403.2 | - | 4 - 5 | 451 | 89 | ||
| JO539083.2 | - | 2 - 3 | 154 | 89 | ||
| JO581235.1 | - | 2 - 3 | 198 | 89 | ||
| JO632237.2 | - | 4 - 5 | 419 | 89 | ||
| JO632238.2 | - | 4 - 5 | 450 | 89 | ||
| JR131732.1 | - | 1 - 2 | 72 | 89 | ||
| JT668209.1 | - | 1 - 2 | 80 | 89 | ||
| JT692850.1 | - | 1 - 2 | 81 | 89 | ||
| JT730034.1 | - | 1 - 2 | 61 | 89 | ||
| JT753906.1 | - | 1 - 2 | 81 | 89 | ||
| KA343066.1 | - | 4 - 5 | 431 | 89 | ||
More Information
Set Membership: |
|

RPL24
Genomic Map