Assay Details
Gene Symbol: SP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675979.3 | XP_005676036.1 | 3 - 4 | 261 | 62 | |
XM_013968933.2 | XP_013824387.1 | 3 - 4 | 618 | 62 | ||
XM_018064451.1 | XP_017919940.1 | 3 - 4 | 543 | 62 | ||
GenBank mRNA | JO401051.2 | - | 3 - 4 | 321 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
