Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678082.3 | XP_005678139.2 | 1 - 2 | 262 | 64 | |
GenBank mRNA | KA306096.1 | - | 1 - 2 | 768 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
