Assay Details
| Gene Symbol: STARD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049977.1 | XP_017905466.1 | 4 - 5 | 381 | 84 | |
| XM_018049979.1 | XP_017905468.1 | 4 - 5 | 399 | 84 | ||
| GenBank mRNA | JO599357.2 | - | 4 - 5 | 361 | 84 | |
More Information
Set Membership: |
|

STARD4
Genomic Map