Assay Details
| Gene Symbol: ALG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687472.3 | XP_005687529.2 | 3 - 4 | 1255 | 58 | |
| XM_018056701.1 | XP_017912190.1 | 3 - 4 | 1255 | 58 | ||
| GenBank mRNA | JO434055.2 | - | 3 - 4 | 1284 | 58 | |
More Information
Set Membership: |
|

ALG11
Genomic Map