Assay Details
Gene Symbol: LOC102187615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060360.1 | XP_017915849.1 | 3 - 4 | 315 | 87 | |
GenBank mRNA | JO585116.2 | - | 3 - 4 | 283 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
