Assay Details
| Gene Symbol: NENF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060749.1 | XP_017916238.1 | 3 - 4 | 410 | 61 | |
| GenBank mRNA | JO376089.2 | - | 3 - 4 | 286 | 61 | |
More Information
Set Membership: |
|

NENF
Genomic Map