Assay Details
Gene Symbol: CNTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038419.1 | XP_017893908.1 | 2 - 3 | 462 | 65 | |
XM_018038420.1 | XP_017893909.1 | 2 - 3 | 384 | 65 | ||
XM_018038421.1 | XP_017893910.1 | 2 - 3 | 462 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
