Assay Details
| Gene Symbol: LOC108635102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045388.1 | XP_017900877.1 | 1249 | 98 | ||
| Gene Symbol: LOC102189920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038506.1 | XP_017893995.1 | 9 - 10 | 1213 | 98 | |
| GenBank mRNA | JO577347.1 | - | 11 - 12 | 1631 | 98 | |
More Information
Set Membership: |
|

LOC108635102
Genomic Map