Assay Details
| Gene Symbol: ERI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697567.3 | XP_005697624.2 | 4 - 5 | 593 | 69 | |
| XM_018040675.1 | XP_017896164.1 | 4 - 5 | 316 | 69 | ||
| XM_018040676.1 | XP_017896165.1 | 4 - 5 | 375 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018040675.1 transcripts.
Set Membership: |
|

ERI2
Genomic Map