Assay Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054121.1 | XP_017909610.1 | 11 - 12 | 2823 | 57 | |
| XM_018054122.1 | XP_017909611.1 | 10 - 11 | 2558 | 57 | ||
| XM_018054123.1 | XP_017909612.1 | 10 - 11 | 1867 | 57 | ||
| GenBank mRNA | JO621145.2 | - | 2 - 3 | 2666 | 57 | |
More Information
Set Membership: |
|

SAMD4A
Genomic Map