Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054631.1 | XP_017910120.1 | 2 - 3 | 474 | 65 | |
| GenBank mRNA | KA336498.1 | - | 4 - 5 | 831 | 65 | |
More Information
Set Membership: |
|

CHMP4A
Genomic Map