Assay Details
| Gene Symbol: C14H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013969044.2 | XP_013824498.1 | 5 - 6 | 611 | 84 | |
| GenBank mRNA | JO587245.2 | - | 4 - 5 | 423 | 84 | |
| JO620379.2 | - | 2 - 3 | 239 | 84 | ||
More Information
Set Membership: |
|

C14H8orf33
Genomic Map