Assay Details
Gene Symbol: MED29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062389.1 | XP_017917878.1 | 3 - 4 | 467 | 71 | |
GenBank mRNA | JO580956.1 | - | 3 - 4 | 383 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
