Assay Details
| Gene Symbol: RAB43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038378.1 | XP_017893867.1 | 2 - 3 | 515 | 58 | |
| XM_018038379.1 | XP_017893868.1 | 2 - 3 | 511 | 58 | ||
| GenBank mRNA | JO413750.2 | - | 2 - 3 | 488 | 58 | |
| JO428402.1 | - | 1 - 2 | 1970 | 58 | ||
More Information
Set Membership: |
|

RAB43
Genomic Map