Assay Details
| Gene Symbol: SCUBE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038776.1 | XP_017894265.1 | 8 - 9 | 1388 | 60 | |
| XM_018038777.1 | XP_017894266.1 | 7 - 8 | 1340 | 60 | ||
| GenBank mRNA | JT674614.1 | - | 9 - 10 | 1258 | 60 | |
| KA327377.1 | - | 14 - 15 | 2059 | 60 | ||
More Information
Set Membership: |
|

SCUBE3
Genomic Map