Assay Details
Gene Symbol: ZNF789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039901.1 | XP_017895390.1 | 2 - 3 | 282 | 74 | |
XM_018039902.1 | XP_017895391.1 | 2 - 3 | 308 | 74 | ||
XM_018039903.1 | XP_017895392.1 | 2 - 3 | 252 | 74 | ||
XM_018039904.1 | XP_017895393.1 | 2 - 3 | 190 | 74 | ||
XM_018039905.1 | XP_017895394.1 | 2 - 3 | 816 | 74 | ||
GenBank mRNA | JO438427.2 | - | 3 - 4 | 939 | 74 | |
KA353247.1 | - | 3 - 4 | 1345 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
