Assay Details
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675884.3 | XP_005675941.1 | 2 - 3 | 445 | 75 | |
GenBank mRNA | KA335585.1 | - | 2 - 3 | 407 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
