Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678363.3 | XP_005678420.1 | 6 - 7 | 760 | 73 | |
| GenBank mRNA | JO577376.1 | - | 6 - 7 | 813 | 73 | |
| JO577377.1 | - | 6 - 7 | 813 | 73 | ||
More Information
Set Membership: |
|

TTC4
Genomic Map