Assay Details
| Gene Symbol: TXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680657.3 | XP_005680714.2 | 2 - 3 | 382 | 83 | |
| GenBank mRNA | JO357513.2 | - | 1 - 2 | 183 | 83 | |
More Information
Set Membership: |
|

TXN2
Genomic Map