Assay Details
Gene Symbol: LARP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053565.1 | XP_017909054.1 | 2 - 3 | 504 | 70 | |
GenBank mRNA | JO633500.2 | - | 2 - 3 | 783 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
