Assay Details
| Gene Symbol: MRPL48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689960.3 | XP_005690017.1 | 3 - 4 | 495 | 65 | |
| GenBank mRNA | JO507203.2 | - | 5 - 6 | 739 | 65 | |
More Information
Set Membership: |
|

MRPL48
Genomic Map