Assay Details
| Gene Symbol: RGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690462.3 | XP_005690519.1 | 2 - 3 | 276 | 82 | |
| GenBank mRNA | JO587397.2 | - | 2 - 3 | 228 | 82 | |
More Information
Set Membership: |
|

RGS1
Genomic Map