Assay Details
| Gene Symbol: SUSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060192.1 | XP_017915681.1 | 4 - 5 | 725 | 64 | |
| XM_018060193.1 | XP_017915682.1 | 4 - 5 | 725 | 64 | ||
| GenBank mRNA | JO636749.2 | - | 6 - 7 | 2213 | 64 | |
More Information
Set Membership: |
|

SUSD4
Genomic Map