Assay Details
| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691996.3 | XP_005692053.1 | 5 - 6 | 631 | 55 | |
| XM_018062009.1 | XP_017917498.1 | 5 - 6 | 631 | 55 | ||
| XM_018062010.1 | XP_017917499.1 | 3 - 4 | 321 | 55 | ||
| GenBank mRNA | KA318510.1 | - | 8 - 9 | 1685 | 55 | |
More Information
Set Membership: |
|

OGFOD1
Genomic Map