Assay Details
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692728.3 | XP_005692785.1 | 5 - 6 | 529 | 71 | |
XM_005692729.3 | XP_005692786.1 | 5 - 6 | 548 | 71 | ||
GenBank mRNA | JO404286.2 | - | 5 - 6 | 626 | 71 | |
JO441929.2 | - | 5 - 6 | 580 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
