Assay Details
Gene Symbol: BPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696908.3 | XP_005696965.1 | 4 - 5 | 801 | 62 | |
XM_018038946.1 | XP_017894435.1 | 6 - 7 | 1028 | 62 | ||
XM_018038947.1 | XP_017894436.1 | 5 - 6 | 787 | 62 | ||
XM_018038948.1 | XP_017894437.1 | 5 - 6 | 1108 | 62 | ||
XM_018038949.1 | XP_017894438.1 | 4 - 5 | 996 | 62 | ||
XM_018038950.1 | XP_017894439.1 | 4 - 5 | 578 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
