Assay Details
| Gene Symbol: MRPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967443.2 | XP_013822897.1 | 2 - 3 | 204 | 77 | |
| XM_013967444.2 | XP_013822898.1 | 2 - 3 | 168 | 77 | ||
| GenBank mRNA | JO544791.2 | - | 4 - 5 | 483 | 77 | |
| KA321068.1 | - | 4 - 5 | 531 | 77 | ||
More Information
Set Membership: |
|

MRPL30
Genomic Map